Methyl 2-chloro-4-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

30671-500MG 30671-1G 30671-5G 30671-25G
77325-128EA 147.29 USD
77325-128 77345-194 77361-466 77384-654
Methyl 2-chloro-4-ethoxybenzoate ≥97%
Methyl 2-chloro-4-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD24368737
CAS Number: 1368274-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR