4-(Benzyloxy)-2,5-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30660-500MG 30660-1G 30660-5G 30660-25G
77325-126EA 263.68 USD
77325-126 77348-908 77361-464 77384-652
4-(Benzyloxy)-2,5-difluorobenzoic acid ≥97%
4-(Benzyloxy)-2,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₃
MW: 264.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503325
CAS Number: 158905-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR