3,5-Dichloro-4-(methoxymethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30650-500MG 30650-1G 30650-5G
77337-832EA 449.08 USD
77337-832 77348-906 77373-170
3,5-Dichloro-4-(methoxymethyl)phenylboronic acid pinacol ester ≥97%
3,5-Dichloro-4-(methoxymethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BCl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30737490
CAS Number: 2121512-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR