Benzyl 2-isopropoxynicotinate ≥97%
Supplier: AOBChem USA
|
Warning
|
30598-500MG
30598-1G
30598-5G
30598-25G
77325-124EA
201.88
USD
77325-124
77348-902
77361-462
77384-650
Benzyl 2-isopropoxynicotinate ≥97%
Benzyl 2-isopropoxynicotinate
Precaución: for Research Use Only
Formula:
C₁₆H₁₇NO₃ MW: 271.311 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29088169 CAS Number: 1826110-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...