2,3-Dimethoxy-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30595-500MG 30595-1G 30595-5G 30595-25G
77325-122EA 284.28 USD
77325-122 77348-900 77361-460 77384-648
2,3-Dimethoxy-4-fluorophenylboronic acid pinacol ester ≥97%
2,3-Dimethoxy-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BFO₄
MW: 282.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088168
CAS Number: 2121512-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR