2-Bromo-4-phenylthiophene ≥97%

Supplier: AOBChem USA

Warning

30588-500MG 30588-1G 30588-5G 30588-25G
77325-120EA 114.33 USD
77325-120 77343-382 77361-458 77373-166
2-Bromo-4-phenylthiophene ≥97%
2-Bromo-4-phenylthiophene

Precaución: for Research Use Only

Formula: C₁₀H₇BrS
MW: 239.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01927272
CAS Number: 10353-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR