4-Cyano-2,5-difluorothioanisole ≥95%

Supplier: AOBChem USA

Warning

30586-500MG 30586-1G 30586-5G 30586-25G
77325-118EA 181.28 USD
77325-118 77346-796 77361-456 77384-646
4-Cyano-2,5-difluorothioanisole ≥95%
4-Cyano-2,5-difluorothioanisole

Precaución: for Research Use Only

Formula: C₈H₅F₂NS
MW: 185.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28765128
CAS Number: 1804896-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR