2-Chloro-5-fluoro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30558-500MG 30558-1G 30558-5G 30558-25G
77325-114EA 126.69 USD
77325-114 77344-202 77361-452 77373-164
2-Chloro-5-fluoro-3-methylbenzyl alcohol ≥97%
2-Chloro-5-fluoro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751965
CAS Number: 1807237-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR