4-Benzyloxy-3,5-dichlorophenylboronic acid pinacol ester ≥95%
Supplier: AOBChem USA
|
Warning
|
30511-500MG
30511-1G
30511-5G
30511-25G
77325-110EA
189.52
USD
77325-110
77348-898
77361-446
77384-640
4-Benzyloxy-3,5-dichlorophenylboronic acid pinacol ester ≥95%
4-Benzyloxy-3,5-dichlorophenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₉H₂₁BCl₂O₃ MW: 379.085 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24039736 CAS Number: 2121512-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...