5-Bromo-2-iodo-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
30463-500MG
30463-1G
30463-5G
30463-25G
77325-108EA
268.83
USD
77325-108
77348-896
77361-444
77384-638
5-Bromo-2-iodo-1,1'-biphenyl ≥97%
5-Bromo-2-iodo-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₂H₈BrI MW: 359 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00091613 CAS Number: 4510-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...