2,6-Difluoro-3-iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

30427-1G 30427-5G 30427-25G
77341-688EA 100.94 USD
77341-688 77361-438 77373-158
2,6-Difluoro-3-iodophenylboronic acid ≥97%
2,6-Difluoro-3-iodophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD32707000
CAS Number: 1447827-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR