Methyl 4-Chloro-2,3-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

30360-500MG 30360-1G 30360-5G
77325-100EA 346.08 USD
77325-100 77348-894 77373-156
Methyl 4-Chloro-2,3-dimethylbenzoate ≥97%
Methyl 4-Chloro-2,3-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088157
CAS Number: 5628-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR