4-Bromo-2,3-dimethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30351-500MG 30351-1G 30351-5G 30351-25G
77325-098EA 156.56 USD
77325-098 77345-600 77361-432 77384-630
4-Bromo-2,3-dimethylphenylboronic acid pinacol ester ≥97%
4-Bromo-2,3-dimethylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BBrO₂
MW: 311.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29036581
CAS Number: 2121511-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR