5-Chloro-2-isopropoxy-pyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

30336-500MG 30336-1G 30336-5G 30336-25G
77325-096EA 243.08 USD
77325-096 77348-886 77361-430 77384-628
5-Chloro-2-isopropoxy-pyridine-3-carbaldehyde ≥97%
5-Chloro-2-isopropoxy-pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28970139
CAS Number: 1289100-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR