4,7-Epoxyisobenzofuran-1,3-dione, 5,6-dibromohexahydro- ≥95%

Supplier: AOBChem USA

Warning

30297-500MG 30297-1G 30297-5G 30297-25G
77325-094EA 95.79 USD
77325-094 77342-538 77361-428 77373-146
4,7-Epoxyisobenzofuran-1,3-dione, 5,6-dibromohexahydro- ≥95%
4,7-Epoxyisobenzofuran-1,3-dione, 5,6-dibromohexahydro-

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₄
MW: 325.939 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464408
CAS Number: 51371-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR