4-(2,4-Difluorophenyl)acetophenone ≥97%

Supplier: AOBChem USA

Warning

30278-500MG 30278-1G 30278-5G
77325-088EA 341.96 USD
77325-088 77348-878 77373-142
4-(2,4-Difluorophenyl)acetophenone ≥97%
4-(2,4-Difluorophenyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O
MW: 232.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907800
CAS Number: 53591-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR