Methyl 2-methyl-4-propan-2-yloxybenzoate ≥97%

Supplier: AOBChem USA

Warning

30272-500MG 30272-1G 30272-5G 30272-25G
77325-086EA 201.88 USD
77325-086 77348-876 77361-424 77384-622
Methyl 2-methyl-4-propan-2-yloxybenzoate ≥97%
Methyl 2-methyl-4-propan-2-yloxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23115088
CAS Number: 175153-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR