4-Bromo-2,6-dimethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30263-500MG 30263-1G 30263-5G 30263-25G
77325-082EA 188.49 USD
77325-082 77348-872 77361-420 77384-620
4-Bromo-2,6-dimethylphenylboronic acid pinacol ester ≥97%
4-Bromo-2,6-dimethylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD29088150
CAS Number: 2121511-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR