3,5-Dibromo-2-(difluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

30262-500MG 30262-1G 30262-5G
77325-080EA 318.27 USD
77325-080 77348-870 77373-136
3,5-Dibromo-2-(difluoromethyl)pyridine ≥97%
3,5-Dibromo-2-(difluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃Br₂F₂N
Storage Temperature: Ambient
MDL Number: MFCD25476744
CAS Number: 1627906-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR