5-Bromo-2-ethoxypyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30227-500MG 30227-1G 30227-5G 30227-25G
77325-078EA 152.44 USD
77325-078 77345-446 77361-418 77384-618
5-Bromo-2-ethoxypyridine-3-boronic acid pinacol ester ≥97%
5-Bromo-2-ethoxypyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BBrNO₃
Storage Temperature: Ambient
MDL Number: MFCD29088144
CAS Number: 2121514-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR