(5-Isopropoxypyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

30216-500MG 30216-1G 30216-25G 30216-5G
77325-076EA 304.88 USD
77325-076 77348-868 77384-616 77373-134
(5-Isopropoxypyridin-3-yl)methanol ≥97%
(5-Isopropoxypyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD22199936
CAS Number: 1546841-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR