4-Bromo-N,N-dimethyl-3-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

30174-1G 30174-5G 30174-25G
77341-382EA 83.43 USD
77341-382 77361-414 77373-132
4-Bromo-N,N-dimethyl-3-(trifluoromethyl)aniline ≥97%
4-Bromo-N,N-dimethyl-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₉H₉BrF₃N
MW: 268.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00219439
CAS Number: 51332-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR