2,5-Diflouro-4-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

30173-500MG 30173-1G 30173-5G 30173-25G
77325-072EA 119.48 USD
77325-072 77343-580 77361-412 77373-130
2,5-Diflouro-4-(methylsulfanyl)benzoic acid ≥97%
2,5-Diflouro-4-(methylsulfanyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂S
MW: 204.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24498146
CAS Number: 918967-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR