Ethyl 3-bromo-5-(trifluoromethoxy)benzoate ≥95%
Supplier: AOBChem USA
|
Warning
|
30132-500MG
30132-1G
30132-5G
30132-25G
77325-068EA
182.31
USD
77325-068
77346-906
77361-408
77384-610
Ethyl 3-bromo-5-(trifluoromethoxy)benzoate ≥95%
Ethyl 3-bromo-5-(trifluoromethoxy)benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₈BrF₃O₃ MW: 313.068 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09999510 CAS Number: 1229442-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...