1-(5-Bromobenzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

30107-500MG 30107-1G 30107-5G
77340-196EA 827.09 USD
77340-196 77348-860 77373-128
1-(5-Bromobenzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone ≥95%
1-(5-Bromobenzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₄BrF₃OS
Storage Temperature: Ambient
MDL Number: MFCD22681647
CAS Number: 1391766-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR