5-Isopropoxy-2-methylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30076-500MG 30076-1G 30076-5G 30076-25G
77325-060EA 112.27 USD
77325-060 77343-328 77361-400 77373-126
5-Isopropoxy-2-methylphenylboronic acid, pinacol ester ≥97%
5-Isopropoxy-2-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₃
MW: 276.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27977091
CAS Number: 1416723-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR