5-Bromo-2-chloro-1-methylpyridin-4(1H)-one ≥97%

Supplier: AOBChem USA

Warning

30067-500MG 30067-1G 30067-5G 30067-25G
77325-058EA 181.28 USD
77325-058 77346-792 77361-398 77384-602
5-Bromo-2-chloro-1-methylpyridin-4(1H)-one ≥97%
5-Bromo-2-chloro-1-methylpyridin-4(1H)-one

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD29036616
CAS Number: 1809157-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR