2-Methyl-1-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

29938-1G 29938-5G 29938-25G
77340-888EA 62.83 USD
77340-888 77361-386 77373-116
2-Methyl-1-naphthoic acid ≥97%
2-Methyl-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00089618
CAS Number: 1575-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR