(((4-bromo-1,3-phenylene)bis(oxy))bis(methylene))dibenzene ≥95%

Supplier: AOBChem USA

Warning

29868-500MG 29868-1G 29868-5G 29868-25G
77325-050EA 99.91 USD
77325-050 77342-764 77361-384 77373-114
(((4-bromo-1,3-phenylene)bis(oxy))bis(methylene))dibenzene ≥95%
(((4-bromo-1,3-phenylene)bis(oxy))bis(methylene))dibenzene

Precaución: for Research Use Only

Formula: C₂₀H₁₇BrO₂
MW: 369.252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00470233
CAS Number: 55583-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR