1,3-Dibromo-5-fluoro-2-methoxy-4-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

29711-1G 29711-5G 29711-25G
77341-266EA 80.34 USD
77341-266 77361-380 77373-112
1,3-Dibromo-5-fluoro-2-methoxy-4-nitrobenzene ≥97%
1,3-Dibromo-5-fluoro-2-methoxy-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₂FNO₃
Storage Temperature: Ambient
MDL Number: MFCD26395361
CAS Number: 346664-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR