Methyl 4-chloro-3-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

29643-500MG 29643-1G 29643-5G 29643-25G
77325-042EA 284.28 USD
77325-042 77348-850 77361-374 77384-590
Methyl 4-chloro-3-isopropylbenzoate ≥97%
Methyl 4-chloro-3-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
MW: 212.673 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29036596
CAS Number: 1809168-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR