2,2-Difluoroethyl methanesulfonate ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,2-Difluoroethylmethane sulphonate, 2,2-Difluoroethyl mesylate
29636-500MG
29636-1G
29636-5G
29636-25G
77325-040EA
165.83
USD
77325-040
77346-170
77361-372
77384-588
2,2-Difluoroethyl methanesulfonate ≥95%
2,2-Difluoroethyl methanesulfonate
Precaución: for Research Use Only
Formula:
C₃H₆F₂O₃S MW: 160.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19442174 CAS Number: 163035-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...