4-[2-(4-iodophenoxy)ethyl]morpholine ≥97%

Supplier: AOBChem USA

Warning

29617-500MG 29617-1G 29617-5G 29617-25G
77325-036EA 161.71 USD
77325-036 77345-832 77361-368 77384-584
4-[2-(4-iodophenoxy)ethyl]morpholine ≥97%
4-[2-(4-iodophenoxy)ethyl]morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₆INO₂
MW: 333.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14646678
CAS Number: 103808-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR