3-Chloro-5-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

29588-500MG 29588-1G 29588-5G 29588-25G
77325-032EA 206 USD
77325-032 77348-846 77361-364 77384-580
3-Chloro-5-(methylthio)pyridine ≥95%
3-Chloro-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNS
MW: 159.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28516422
CAS Number: 98627-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR