5-Bromo-2,3-difluoro-4-(trimethylsilyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

29578-500MG 29578-1G 29578-5G 29578-25G
77325-030EA 140.08 USD
77325-030 77344-690 77361-362 77384-578
5-Bromo-2,3-difluoro-4-(trimethylsilyl)benzoic acid ≥95%
5-Bromo-2,3-difluoro-4-(trimethylsilyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrF₂O₂Si
MW: 309.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32706999
CAS Number: 1241948-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR