4-Isopropoxy-3,5-dimethyl-benzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

29575-500MG 29575-1G 29575-5G 29575-25G
77325-028EA 263.68 USD
77325-028 77348-844 77361-358 77384-576
4-Isopropoxy-3,5-dimethyl-benzoic acid methyl ester ≥97%
4-Isopropoxy-3,5-dimethyl-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24038455
CAS Number: 1429901-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR