4-Isopropoxy-3-methoxyphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29570-500MG 29570-1G 29570-5G
77325-026EA 346.08 USD
77325-026 77348-842 77373-108
4-Isopropoxy-3-methoxyphenylboronic acid, pinacol ester ≥97%
4-Isopropoxy-3-methoxyphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₄
MW: 292.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22495044
CAS Number: 1163700-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR