2'-Fluoro-4'-methyl-[1,1'-biphenyl]-4-ol ≥95%

Supplier: AOBChem USA

Warning

29565-500MG 29565-1G 29565-5G
77325-024EA 347.11 USD
77325-024 77348-840 77373-106
2'-Fluoro-4'-methyl-[1,1'-biphenyl]-4-ol ≥95%
2'-Fluoro-4'-methyl-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO
MW: 202.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18312798
CAS Number: 1261911-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR