1-Bromo-4-isopropoxy-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

29525-500MG 29525-1G 29525-5G 29525-25G
77325-020EA 196.73 USD
77325-020 77348-836 77361-354 77384-572
1-Bromo-4-isopropoxy-2-methoxybenzene ≥97%
1-Bromo-4-isopropoxy-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27946224
CAS Number: 176391-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR