2-Isopropoxy-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

29521-500MG 29521-1G 29521-5G
77336-992EA 403.76 USD
77336-992 77348-834 77373-104
2-Isopropoxy-4-methylbenzoic acid ≥95%
2-Isopropoxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14652964
CAS Number: 689161-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR