6-Bromo-5-methoxypyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

29389-500MG 29389-1G 29389-5G
77337-830EA 449.08 USD
77337-830 77348-830 77373-100
6-Bromo-5-methoxypyridin-3-ol ≥97%
6-Bromo-5-methoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂
MW: 204.021 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257134
CAS Number: 1256833-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR