2-Bromo-3-methoxypyridine-5-boronic acid ≥95%

Supplier: AOBChem USA

Warning

29387-500MG 29387-1G 29387-5G
77336-774EA 394.49 USD
77336-774 77348-828 77373-098
2-Bromo-3-methoxypyridine-5-boronic acid ≥95%
2-Bromo-3-methoxypyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BBrNO₃
Storage Temperature: Ambient
MDL Number: MFCD22491366
CAS Number: 2121512-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR