2-Bromo-4-chloro-1-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

29344-500MG 29344-1G 29344-5G 29344-25G
77325-016EA 192.61 USD
77325-016 77348-824 77361-352 77384-570
2-Bromo-4-chloro-1-isopropylbenzene ≥95%
2-Bromo-4-chloro-1-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrCl
MW: 233.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356419
CAS Number: 1369828-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR