5-Chloro-2-ethylaniline ≥95%

Supplier: AOBChem USA

Warning

29341-1G 29341-5G 29341-25G
77341-640EA 97.85 USD
77341-640 77361-350 77373-094
5-Chloro-2-ethylaniline ≥95%
5-Chloro-2-ethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20694404
CAS Number: 3843-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR