5-Chloro-2-(1-methylethyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
29335-500MG
29335-1G
29335-5G
77340-150EA
796.19
USD
77340-150
77348-822
77373-092
5-Chloro-2-(1-methylethyl)phenylboronic acid ≥97%
5-Chloro-2-(1-methylethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₂BClO₂ MW: 198.454 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23380981 CAS Number: 1403679-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...