1-Fluoro-3-isopropoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

29321-1G 29321-5G 29321-25G
77340-622EA 60.77 USD
77340-622 77361-348 77373-090
1-Fluoro-3-isopropoxy-5-methylbenzene ≥97%
1-Fluoro-3-isopropoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114435
CAS Number: 1369853-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR