1-Fluoro-3-ethoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

29319-1G 29319-5G 29319-25G
77341-392EA 84.46 USD
77341-392 77361-346 77373-088
1-Fluoro-3-ethoxy-5-methylbenzene ≥97%
1-Fluoro-3-ethoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806034
CAS Number: 1809158-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR