Ethyl 3,5-dichloro-4-propan-2-yloxybenzoate ≥97%

Supplier: AOBChem USA

Warning

29304-500MG 29304-1G 29304-5G 29304-25G
77325-014EA 173.04 USD
77325-014 77346-438 77361-344 77384-568
Ethyl 3,5-dichloro-4-propan-2-yloxybenzoate ≥97%
Ethyl 3,5-dichloro-4-propan-2-yloxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂O₃
MW: 277.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088129
CAS Number: 40689-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR