2,5-Difluoro-4-(methylsulfanyl)phenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29294-500MG 29294-1G 29294-5G 29294-25G
77325-010EA 120.51 USD
77325-010 77343-674 77361-342 77373-084
2,5-Difluoro-4-(methylsulfanyl)phenylboronic acid, pinacol ester ≥97%
2,5-Difluoro-4-(methylsulfanyl)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD23380285
CAS Number: 1383968-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR