3,5-Dichloro-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29290-500MG 29290-1G 29290-5G 29290-25G
77325-008EA 94.76 USD
77325-008 77342-522 77361-340 77373-082
3,5-Dichloro-4-isopropoxybenzoic acid ≥97%
3,5-Dichloro-4-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₃
MW: 249.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24038824
CAS Number: 41490-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR